Trimethylsilyl methacrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: stabiliseret med BHT/Ionol

T3293-5ML T3293-25ML
TCIAT3293-5MLEA 238 DKK
TCIAT3293-5ML TCIAT3293-25ML
Trimethylsilyl methacrylate ≥98.0% (by GC) stabilised
Trimethylsilyl methacrylate
Formel: C₇H₁₄O₂Si
Kogepunkt: 52°C
Flammepunkt: 32°C
MDL Number: MFCD00053868
CAS-nummer: 13688-56-7
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=418fdb9ad43334c3b4cc96d63c05ded5983dfed4&vid=418fdb9ad43334c3b4cc96d63c05ded5983dfed4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR