2,3,4-Trimethoxybenzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T3285-25G
T3285-5G
TCIAT3285-25GEA
1690
DKK
TCIAT3285-25G
TCIAT3285-5G
2,3,4-Trimethoxybenzonitril ≥98.0% (by GC)
2,3,4-Trimethoxybenzonitril
Formel:
C₁₀H₁₁NO₃ Kogepunkt: 167°C |
MDL Number:
MFCD00001785 CAS-nummer: 43020-38-8 EINECS: 256-049-5 UN nummer: 3439 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...