2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-(4-Trifluoromethylphenyl)ethylamine, 4-(Trifluoromethyl)phenethylamine

T3278-1G T3278-5G
TCIAT3278-1GEA 247 DKK
TCIAT3278-1G TCIAT3278-5G
2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)
2-[4-(Trifluoromethyl)phenyl]ethylamine
Formel: C₉H₁₀F₃N
Kogepunkt: 90°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01529875
CAS-nummer: 775-00-8
UN nummer: 2735
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR