Triphenylen ≥98.0% (by GC), purified by sublimation
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
9,10-Benzophenanthrene, Isochrysene
T3267-200MG
T3267-1G
TCIAT3267-200MGEA
526
DKK
TCIAT3267-200MG
TCIAT3267-1G
Triphenylen ≥98.0% (by GC), purified by sublimation
Triphenylene
Formel:
C₁₈H₁₂ Molekylvægt: 228.29 g/mol Kogepunkt: 425°C Smeltepunkt: 199 |
MDL Number:
MFCD00001108 CAS-nummer: 217-59-4 EINECS: 205-922-9 Merck Index: 14,09740 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 197.0 to 201.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...