Ethylentetracarbonitril ≥98.0% (by total nitrogen basis), purified by sublimation

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T3264-1G T3264-5G
TCIAT3264-1GEA 427 DKK
TCIAT3264-1G TCIAT3264-5G
Ethylentetracarbonitril ≥98.0% (by total nitrogen basis), purified by sublimation
Ethylentetracarbonitril
Formel: C₆N₄
Kogepunkt: 223°C
MDL Number: MFCD00001850
CAS-nummer: 670-54-2
EINECS: 211-578-0
UN nummer: 3439
ADR: 6.1,I
Merck Index: 14,09195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59d528570d1ab8216fcbb540a021816f1b95f6f3&vid=59d528570d1ab8216fcbb540a021816f1b95f6f3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR