Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) ≥95.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T3255-1G
T3255-5G
TCIAT3255-1GEA
1010
DKK
TCIAT3255-1G
TCIAT3255-5G
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) ≥95.0% (by titrimetric analysis)
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate)
Formel:
C₃₀H₂₄CoF₁₂N₆P₂ |
CAS-nummer:
79151-78-3 |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Elemental analysis (Nitrogen) | 9.70 to 10.80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...