3,4,5-Trimethoxybenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T3244-5G T3244-25G
TCIAT3244-5GEA 509 DKK
TCIAT3244-5G TCIAT3244-25G
3,4,5-Trimethoxybenzonitril ≥98.0% (by GC)
3,4,5-Trimethoxybenzonitril
Formel: C₁₀H₁₁NO₃
Kogepunkt: 185°C
MDL Number: MFCD00001803
CAS-nummer: 1885-35-4
EINECS: 217-550-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=069f3ac0ffcf8f637389bf82187d273add4c6d0c&vid=069f3ac0ffcf8f637389bf82187d273add4c6d0c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR