5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Dibenzo[a,e]cyclooctadien-5,11-diyne, 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene

T3241-100MG T3241-500MG
TCIAT3241-100MGEA 1420 DKK
TCIAT3241-100MG TCIAT3241-500MG
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene ≥98.0% (by HPLC)
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene
Formel: C₁₆H₈
Molekylvægt: 200.24 g/mol
Smeltepunkt: 110°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD28386121
CAS-nummer: 53397-65-2

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR