Trimethyl(phenoxy)silane ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Phenyl trimethylsilyl ether

T3222-5ML T3222-25ML
TCIAT3222-5MLEA 206 DKK
TCIAT3222-5ML TCIAT3222-25ML
Trimethyl(phenoxy)silane ≥97.0% (by GC)
Trimethyl(phenoxy)silane
Formel: C₉H₁₄OSi
Molekylvægt: 166.30 g/mol
Kogepunkt: 211°C
Smeltepunkt: –55°C
Flammepunkt: 41°C
MDL Number: MFCD00053666
CAS-nummer: 1529-17-5
EINECS: 216-211-8
UN nummer: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64bd6c9e8f756a2a4c74e103e1b09ca348ec8ceb&vid=64bd6c9e8f756a2a4c74e103e1b09ca348ec8ceb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR