(2-Thienylethynyl)trimethylsilane ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(Trimethylsilylethynyl)thiophene, Trimethyl(thiophen-2-ylethynyl)silane
T3189-1G
T3189-5G
TCIAT3189-1GEA
788
DKK
TCIAT3189-1G
TCIAT3189-5G
(2-Thienylethynyl)trimethylsilane ≥98.0% (by GC)
(2-Thienylethynyl)trimethylsilane
Formel:
C₉H₁₂SSi Kogepunkt: 90°C Flammepunkt: 25°C |
MDL Number:
MFCD03427125 CAS-nummer: 40231-03-6 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...