Triethylene glycol butyl methyl ether ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane
T3181-25ML
T3181-500ML
TCIAT3181-25MLEA
132
DKK
TCIAT3181-25ML
TCIAT3181-500ML
Triethylene glycol butyl methyl ether ≥98.0% (by GC)
Triethylene glycol butyl methyl ether
Formel:
C₁₁H₂₄O₄ Molekylvægt: 220.31 g/mol Smeltepunkt: –34°C |
MDL Number:
MFCD23703159 CAS-nummer: 7382-30-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...