(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T3153-1G
T3153-5G
TCIAT3153-1GEA
476
DKK
TCIAT3153-1G
TCIAT3153-5G
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formel:
C₄₈H₄₀P₂ Smeltepunkt: 258°C |
MDL Number:
MFCD01311709 CAS-nummer: 100165-88-6 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 99.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...