2,4,6-Tribromophenyl acrylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acrylic Acid 2,4,6-Tribromophenyl Ester

T3130-5G T3130-25G
TCIAT3130-5GEA 386 DKK
TCIAT3130-5G TCIAT3130-25G
2,4,6-Tribromophenyl acrylate ≥98.0% (by GC)
2,4,6-Tribromophenyl acrylate
Formel: C₉H₅Br₃O₂
MDL Number: MFCD00078430
CAS-nummer: 3741-77-3
EINECS: 223-132-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d031fa067c5cf6ad2363bb54f1654fcff155486&vid=0d031fa067c5cf6ad2363bb54f1654fcff155486&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR