2,4,6-Trichlorophenyl formate ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Formic Acid 2,4,6-Trichlorophenyl Ester, 2,4,6-Trichlorophenyl Formate

T3121-5G T3121-1G
TCIAT3121-5GEA 657 DKK
TCIAT3121-5G TCIAT3121-1G
2,4,6-Trichlorophenyl formate ≥97.0% (by HPLC)
2,4,6-Trichlorophenyl formate
Formel: C₇H₃Cl₃O₂
MDL Number: MFCD27996252
CAS-nummer: 4525-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8a1352773feca3e04b69a6f790a579c765f5419&vid=f8a1352773feca3e04b69a6f790a579c765f5419&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR