1,2,3-Trimethyl-3H-benzo[e]indole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3119-5G T3119-25G
TCIAT3119-5GEA 444 DKK
TCIAT3119-5G TCIAT3119-25G
1,2,3-Trimethyl-3H-benzo[e]indole ≥98.0% (by GC)
1,2,3-Trimethyl-3H-benzo[e]indole
Formel: C₁₅H₁₅N
MDL Number: MFCD06651515
CAS-nummer: 881219-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3432b2f8e97b8ea00740a6c8a32de3a6634a9f7&vid=a3432b2f8e97b8ea00740a6c8a32de3a6634a9f7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR