Thioridazine hydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 10-[2-(1-Methylpiperidin-2-yl)ethyl]-2-(methylsulphanyl)-10H-phenothiazine hydrochloride

T3102-5G T3102-25G
TCIAT3102-5GEA 591 DKK
TCIAT3102-5G TCIAT3102-25G
Thioridazine hydrochloride ≥98.0% (by HPLC)
Thioridazine hydrochloride
Formel: C₂₁H₂₇ClN₂S₂
Molekylvægt: 407.031 g/mol
MDL Number: MFCD00012655
CAS-nummer: 130-61-0
EINECS: 204-992-8
UN nummer: 3077
ADR: 9 III
Merck Index: 14,09359
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b827296eb623fdfe15d8139cb7a540874b0ff93d&vid=b827296eb623fdfe15d8139cb7a540874b0ff93d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR