Talniflumate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Oxo-1,3-dihydroisobenzofuran-1-yl 2-((3-(trifluoromethyl)phenyl)amino)nicotinate

T3101-200MG T3101-1G
TCIAT3101-200MGEA 1290 DKK
TCIAT3101-200MG TCIAT3101-1G
Talniflumate ≥98.0% (by HPLC, total nitrogen)
Talniflumate
Formel: C₂₁H₁₃F₃N₂O₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00866135
CAS-nummer: 66898-62-2
Merck Index: 14,09044
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d66860ef19a2b0fded445a8530f2ed7cfb55b91&vid=7d66860ef19a2b0fded445a8530f2ed7cfb55b91&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR