4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T2997-5G T2997-25G
TCIAT2997-5GEA 156 DKK
TCIAT2997-5G TCIAT2997-25G
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione ≥98.0% (by GC)

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR