Dihydrofuran-3(2H)-one ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T2971-1G
T2971-5G
TCIAT2971-1GEA
345
DKK
TCIAT2971-1G
TCIAT2971-5G
Dihydrofuran-3(2H)-one ≥98.0% (by GC)
Dihydrofuran-3(2H)-one
Formel:
C₄H₆O₂ Kogepunkt: 68°C Flammepunkt: 56°C |
MDL Number:
MFCD07778393 CAS-nummer: 22929-52-8 UN nummer: 1224 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...