1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T2951-1G
T2951-5G
TCIAT2951-1GEA
132
DKK
TCIAT2951-1G
TCIAT2951-5G
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole ≥98.0% (by HPLC)
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole
Formel:
C₁₇H₂₅N₃O₂S |
MDL Number:
MFCD00014089 CAS-nummer: 54230-60-3 EINECS: 259-035-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 12.00 to 13.00 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...