Tetrakis(4-aminophenyl)methane ≥95.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4',4'',4'''-Methanetetrayltetraaniline

T2947-200MG T2947-1G
TCIAT2947-200MGEA 460 DKK
TCIAT2947-200MG TCIAT2947-1G
Tetrakis(4-aminophenyl)methane ≥95.0% (by HPLC, titration analysis)
Tetrakis(4-aminophenyl)methane
Formel: C₂₅H₂₄N₄
Smeltepunkt: 320°C
MDL Number: MFCD12546930
CAS-nummer: 60532-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f279f664bdbb86d9194b543b74e81bce69b1a4a&vid=1f279f664bdbb86d9194b543b74e81bce69b1a4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR