2-(2-Thienyl)pyridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene

T2936-5G T2936-25G
TCIAT2936-5GEA 657 DKK
TCIAT2936-5G TCIAT2936-25G
2-(2-Thienyl)pyridine ≥98.0% (by GC)
2-(2-Thienyl)pyridine
Formel: C₉H₇NS
Kogepunkt: 140°C
MDL Number: MFCD00044441
CAS-nummer: 3319-99-1
EINECS: 222-022-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4fab058f787eb15ec4e710eb45900e1a53517a5&vid=a4fab058f787eb15ec4e710eb45900e1a53517a5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR