Tranilast ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-[[3-(3,4-Dimethoxyphenyl)-1-oxo-2-propenyl]amino]benzoic Acid, 2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid, N-(3,4-Dimethoxycinnamoyl)anthranilic Acid

T2935-1G T2935-5G
TCIAT2935-1GEA 649 DKK
TCIAT2935-1G TCIAT2935-5G
Tranilast ≥98.0% (by HPLC, titration analysis)
Tranilast
Formel: C₁₈H₁₇NO₅
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00864787
CAS-nummer: 53902-12-8
Merck Index: 14,09570
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=758384eb3df25893ed0e6c282aee125a6857ecd7&vid=758384eb3df25893ed0e6c282aee125a6857ecd7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR