2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Tris(2,4,6-tribromophenyl) Cyanurate, Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester

T2871-5G T2871-25G
TCIAT2871-5GEA 214 DKK
TCIAT2871-5G TCIAT2871-25G
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine
Formel: C₂₁H₆Br₉N₃O₃
MDL Number: MFCD03092948
CAS-nummer: 25713-60-4
EINECS: 426-040-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de1dc6b7af44d1977abaec7e2e8a21c67f71461&vid=3de1dc6b7af44d1977abaec7e2e8a21c67f71461&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 228.0 to 232.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR