Telmisartan ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4'-[[4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylic Acid, 4'-((1,7'-Dimethyl-2'-propyl-1H,3'H-[2,5'-bibenzo[d]imidazol]-3'-yl)methyl)-[1,1'-biphenyl]-2-carboxylic acid

T2861-5G T2861-1G
TCIAT2861-5GEA 1510 DKK
TCIAT2861-5G TCIAT2861-1G
Telmisartan ≥98.0% (by HPLC, titration analysis)
Telmisartan
Formel: C₃₃H₃₀N₄O₂
MDL Number: MFCD00918125
CAS-nummer: 144701-48-4
Merck Index: 14,09129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7a086d2d87d727fc812b37c22c4e2f027b55bd6&vid=f7a086d2d87d727fc812b37c22c4e2f027b55bd6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR