Triclabendazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzo[d]imidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole

T2826-1G T2826-5G
TCIAT2826-1GEA 460 DKK
TCIAT2826-1G TCIAT2826-5G
Triclabendazole ≥98.0% (by HPLC, titration analysis)
Triclabendazole
Formel: C₁₄H₉Cl₃N₂OS
MDL Number: MFCD00864519
CAS-nummer: 68786-66-3
Merck Index: 14,09652

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR