Terazosin hydrochloride dihydrate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Amino-6,7-dimethoxy-2-[4-(tetrahydro-2-furoyl)-1-piperazinyl]quinazoline Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(tetrahydrofuran-2-yl)methanone hydrochloride dihydrate

T2751-1G T2751-100MG
TCIAT2751-1GEA 1380 DKK
TCIAT2751-1G TCIAT2751-100MG
Terazosin hydrochloride dihydrate ≥98.0% (by HPLC, total nitrogen)
Terazosin hydrochloride dihydrate
Formel: C₁₉H₃₀ClN₅O₆
Smeltepunkt: 274°C
MDL Number: MFCD00941408
CAS-nummer: 70024-40-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water 7.0 to 9.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR