Tenoxicam ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide
T2750-25G
T2750-5G
TCIAT2750-25GEA
1790
DKK
TCIAT2750-25G
TCIAT2750-5G
Tenoxicam ≥98.0% (by HPLC, total nitrogen)
Tenoxicam
Formel:
C₁₃H₁₁N₃O₄S₂ Smeltepunkt: 213°C |
MDL Number:
MFCD00083502 CAS-nummer: 59804-37-4 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,09148 |
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...