2,3,5,6-Tetrafluoro-1,4-benzenedimethanol ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha'-diol, 2,3,5,6-Tetrafluoro-alpha,alpha'-dihydroxy-p-xylene
T2732-25G
T2732-5G
TCIAT2732-25GEA
1450
DKK
TCIAT2732-25G
TCIAT2732-5G
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol ≥97.0% (by GC)
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol
Formel:
C₈H₆F₄O₂ |
MDL Number:
MFCD00229147 CAS-nummer: 92339-07-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...