Tetrakis(acetonitrile)copper(I) tetrafluoroborate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Copper(I) tetra(acetonitrile) tetrafluoroborate, Tetrakis(acetonitrile)copper(I) tetrafluoroborate
T2666-1G
T2666-5G
T2666-25G
TCIAT2666-1GEA
165
DKK
TCIAT2666-1G
TCIAT2666-5G
TCIAT2666-25G
Tetrakis(acetonitrile)copper(I) tetrafluoroborate ≥98.0% (by titrimetric analysis)
Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Formel:
C₈H₁₂BCuF₄N₄ |
MDL Number:
MFCD09265110 CAS-nummer: 15418-29-8 UN nummer: 1759 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Potassium permanganate method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...