Thiopicolinamide ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Pyridine-2-carbothioamide, 2-Pyridinethioamide, 2-Pyridylthiocarboxamide

T2618-1G T2618-5G
TCIAT2618-1GEA 165 DKK
TCIAT2618-1G TCIAT2618-5G
Thiopicolinamide ≥97.0% (by GC, titration analysis)
Thiopicolinamide
Formel: C₆H₆N₂S
Molekylvægt: 138.19 g/mol
MDL Number: MFCD00087576
CAS-nummer: 5346-38-3

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR