Dihydroxybis(hydrogen Lactato)titanium(IV) ∼44% in 2-propanol/water

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Titanium(IV) bis(hydrogen Lactato)dihydroxide

T2612-25G T2612-500G
TCIAT2612-25GEA 132 DKK
TCIAT2612-25G TCIAT2612-500G
Dihydroxybis(hydrogen Lactato)titanium(IV) ∼44% in 2-propanol/water
Dihydroxybis(hydrogen Lactato)titanium(IV)
Formel: C₆H₁₂O₈Ti
Molekylvægt: 260.02 g/mol
CAS-nummer: 79110-90-0
UN nummer: 2920
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white clear liquid
Concentration (Gravimetric Method) 42.0 to 46.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR