Dihydroxybis(ammonium lactato)titanium(IV) ∼40% 2-propanol/water
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T2611-25G
T2611-500G
TCIAT2611-25GEA
140
DKK
TCIAT2611-25G
TCIAT2611-500G
Dihydroxybis(ammonium lactato)titanium(IV) ∼40% 2-propanol/water
Dihydroxybis(ammonium lactato)titanium(IV)
Formel:
C₆H₂₂N₂O₈Ti Molekylvægt: 298.115 g/mol |
MDL Number:
MFCD00072736 CAS-nummer: 65104-06-5 EINECS: 265-409-0 UN nummer: 1219 ADR: 3 II |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Concentration (Gravimetric Method) | 37.0 to 45.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...