Triisopropoxyvanadium(V) oxide ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Vanadium(V) Triisopropoxide Oxide, Vanadium (V) tri-i-propoxy oxide, Vanadium(V) Oxytriisopropoxide, VTIP
T2548-25G
T2548-5G
TCIAT2548-25GEA
1150
DKK
TCIAT2548-25G
TCIAT2548-5G
Triisopropoxyvanadium(V) oxide ≥97.0% (by titrimetric analysis)
Triisopropoxyvanadium(V) oxide
Formel:
C₉H₂₁O₄V Molekylvægt: 244.20 g/mol Kogepunkt: 102°C Smeltepunkt: –2°C Massefylde: 1.04 Flammepunkt: 70°C |
MDL Number:
MFCD00015017 CAS-nummer: 5588-84-1 EINECS: 226-997-4 |
Specifikationer Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...