Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tris(3,5-bis(trifluoromethyl)phenyl)phosphine

T2526-1G T2526-5G
TCIAT2526-1GEA 1390 DKK
TCIAT2526-1G TCIAT2526-5G
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98.0% (by GC)
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Formel: C₂₄H₉F₁₈P
Kogepunkt: 130°C
MDL Number: MFCD00042428
CAS-nummer: 175136-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53308729102d6e231d19b37816eb7859a526f350&vid=53308729102d6e231d19b37816eb7859a526f350&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR