(+/-)-Thalidomide ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T2524-1G
T2524-5G
TCIAT2524-1GEA
575
DKK
TCIAT2524-1G
TCIAT2524-5G
(+/-)-Thalidomide ≥98.0% (by HPLC, total nitrogen)
(+/-)-Thalidomide
Formel:
C₁₃H₁₀N₂O₄ |
MDL Number:
MFCD00153873 CAS-nummer: 50-35-1 EINECS: 200-031-1 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,09255 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...