Trimethyl-1,2,3-propanetricarboxylate ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T2429-1G
T2429-5G
TCIAT2429-1GEA
345
DKK
TCIAT2429-1G
TCIAT2429-5G
Trimethyl-1,2,3-propanetricarboxylate ≥97.0% (by GC)
Trimethyl-1,2,3-propanetricarboxylate
Formel:
C₉H₁₄O₆ Kogepunkt: 150°C Massefylde: 1,18 |
MDL Number:
MFCD00040552 CAS-nummer: 6138-26-7 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...