2-(p-Toluenesulphonyl)acetic acid ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T2416-25G
T2416-5G
TCIAT2416-25GEA
2700
DKK
TCIAT2416-25G
TCIAT2416-5G
2-(p-Toluenesulphonyl)acetic acid ≥98.0% (by HPLC)
2-(p-Toluenesulphonyl)acetic acid
Formel:
C₉H₁₀O₄S |
MDL Number:
MFCD00021764 CAS-nummer: 3937-96-0 EINECS: 223-518-0 |
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...