Tetrahydrofurfuryl acrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (Tetrahydrofuran-2-yl)methyl acrylate, Acrylic Acid Tetrahydrofurfuryl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T2239-25G T2239-500G
TCIAT2239-25GEA 140 DKK
TCIAT2239-25G TCIAT2239-500G
Tetrahydrofurfuryl acrylate ≥98.0% (by GC) stabilised
Tetrahydrofurfuryl acrylate
Formel: C₈H₁₂O₃
Molekylvægt: 156.18 g/mol
Kogepunkt: 87°C
Flammepunkt: 95°C
MDL Number: MFCD00014516
CAS-nummer: 2399-48-6
EINECS: 219-268-7

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR