N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate

T2224-1G T2224-5G
TCIAT2224-1GEA 132 DKK
TCIAT2224-1G TCIAT2224-5G
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate
Formel: C₉H₁₆BF₄N₃O₃
Molekylvægt: 301.05 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00077875
CAS-nummer: 105832-38-0
UN nummer: 1759
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR