Sulphur trioxide triethylamine complex ≥96.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T2136-25G T2136-5G
TCIAT2136-25GEA 1100 DKK
TCIAT2136-25G TCIAT2136-5G
Sulphur trioxide triethylamine complex ≥96.0% (by total nitrogen basis)
Sulphur trioxide triethylamine complex
Formel: C₆H₁₅NO₃S
Smeltepunkt: 93°C
Opbevaringstemperatur: Køleskab
CAS-nummer: 761-01-3
UN nummer: 3261
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR