(S)-(+)-p-Toluenesulphinamide ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-4-Methylbenzenesulphinamide

T2069-1G T2069-5G
TCIAT2069-1GEA 649 DKK
TCIAT2069-1G TCIAT2069-5G
(S)-(+)-p-Toluenesulphinamide ≥98.0% (by HPLC)
(S)-(+)-p-Toluenesulphinamide
Formel: C₇H₉NOS
MDL Number: MFCD06858375
CAS-nummer: 188447-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0988bdbec14f33d4be5ebf53950055d3a5583c1a&vid=0988bdbec14f33d4be5ebf53950055d3a5583c1a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 118.0 to 122.0 deg-C
Specific rotation [a]20/D +82.0 to +87.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR