3,5,6-Trifluoro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T2061-1G T2061-5G
TCIAT2061-1GEA 567 DKK
TCIAT2061-1G TCIAT2061-5G
3,5,6-Trifluoro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
3,5,6-Trifluoro-2-hydroxypyridine
Formel: C₅H₂F₃NO
MDL Number: MFCD04114299
CAS-nummer: 75777-49-0

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 102.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR