Triethylsilanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydroxytriethylsilane, Triethyl(hydroxy)silane

T2017-25ML T2017-5ML
TCIAT2017-25MLEA 2150 DKK
TCIAT2017-25ML TCIAT2017-5ML
Triethylsilanol ≥98.0% (by GC)
Triethylsilanol
Formel: C₆H₁₆OSi
Molekylvægt: 132.28 g/mol
MDL Number: MFCD00042640
CAS-nummer: 597-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbb6cc8274aa40c7dd217a0309180c2964a84fe7&vid=bbb6cc8274aa40c7dd217a0309180c2964a84fe7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR