(Triphenylphosphoranylidene)acetaldehyde ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane

T2001-5G T2001-25G
TCIAT2001-5GEA 452 DKK
TCIAT2001-5G TCIAT2001-25G
(Triphenylphosphoranylidene)acetaldehyde ≥98.0% (by HPLC)
(Triphenylphosphoranylidene)acetaldehyde
Formel: C₂₀H₁₇OP
Smeltepunkt: 198°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006994
CAS-nummer: 2136-75-6
EINECS: 218-375-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e48f37235609ce09dc0aebab7fadc0214183952&vid=6e48f37235609ce09dc0aebab7fadc0214183952&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR