Triethylene glycol monobenzyl ether ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol, 2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethanol

T1996-25G T1996-5G
TCIAT1996-25GEA 4140 DKK
TCIAT1996-25G TCIAT1996-5G
Triethylene glycol monobenzyl ether ≥97.0% (by GC)
Triethylene glycol monobenzyl ether
Formel: C₁₃H₂₀O₄
Molekylvægt: 240.30 g/mol
MDL Number: MFCD02093556
CAS-nummer: 55489-58-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR