1,4,8,11-Tetrathiacyclotetradecane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1959-1G T1959-200MG
TCIAT1959-1GEA 4030 DKK
TCIAT1959-1G TCIAT1959-200MG
1,4,8,11-Tetrathiacyclotetradecane ≥98.0% (by GC)
1,4,8,11-Tetrathiacyclotetradecane
Formel: C₁₀H₂₀S₄
MDL Number: MFCD00010048
CAS-nummer: 24194-61-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR