(Triphenylphosphoranylidene)acetonitrile ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T1958-25G T1958-5G
TCIAT1958-25GEA 1260 DKK
TCIAT1958-25G TCIAT1958-5G
(Triphenylphosphoranylidene)acetonitrile ≥98.0% (by HPLC, total nitrogen)
(Triphenylphosphoranylidene)acetonitrile
Formel: C₂₀H₁₆NP
MDL Number: MFCD00567633
CAS-nummer: 16640-68-9
EINECS: 240-689-7
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 190.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR