Terephthalamic acid ≥95.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Terephthalamidic Acid, Terephthalic acid monoamide

T1768-25G T1768-5G
TCIAT1768-25GEA 4650 DKK
TCIAT1768-25G TCIAT1768-5G
Terephthalamic acid ≥95.0% (by HPLC)
Terephthalamic acid
Formel: C₈H₇NO₃
Smeltepunkt: 248°C
MDL Number: MFCD00025482
CAS-nummer: 6051-43-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR