4-(p-Tolyl)butyric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1727-1G T1727-25G T1727-5G
TCIAT1727-1GEA 148 DKK
TCIAT1727-1G TCIAT1727-25G TCIAT1727-5G
4-(p-Tolyl)butyric acid ≥98.0% (by GC, titration analysis)
4-(p-Tolyl)butyric acid
Formel: C₁₁H₁₄O₂
Kogepunkt: 150°C
MDL Number: MFCD00021786
CAS-nummer: 4521-22-6
EINECS: 224-848-8

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR